missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Powder, spherical, 99% (Metals basis), Alfa Aesar™

Brand:  Alfa Aesar 010576.22

Code : NEW

Additional Details : CAS Number : 7429-90-5 Weight : 0.10000kg Transport : UN number : 1396 Chem class : 1 Pack group : II


Quantity 100g

 View more versions of this product

Product Code. 15405955

  • £14.40 / 100g


Estimated Shipment date
from Supplier 26-03-2018
Add to basket


Specifications

Chemical Identifiers

Aluminum
Spherical Powder
MFCD00134029
7429-90-5
99% (Metals basis)

Specifications

Aluminum
7429-90-5
2.7g/cm3
Spherical Powder
MFCD00134029
Silver-gray
100g
Silver Gray powder
400°C (752°F)
99% (Metals basis)
Odorless
Videos
SDS