missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Powder, -40+325 mesh, 99.8% (Metals basis), Alfa Aesar™

Brand:  Alfa Aesar 000010.22

Code : NEW

Additional Details : CAS Number : 7429-90-5 Weight : 0.10000kg Transport : UN number : 1396 Chem class : 1 Pack group : II


Quantity 100g

 View more versions of this product

Product Code. 15425895

  • £17.50 / 100g


Estimated Shipment date
from Supplier 20-02-2018
Add to basket


Specifications

Chemical Identifiers

Aluminum
Powder
MFCD00134029
7429-90-5
99.8% (Metals basis)

Specifications

Aluminum
7429-90-5
2.7g/cm3
Powder
MFCD00134029
Moisture sensitive
100g
Silver Gray powder
400°C (752°F)
99.8% (Metals basis)
Odorless
Silver-gray
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Aluminum powder

Signal Word
  • Danger

Hazard Category
  • Pyrophoric solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 2

Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H261-In contact with water releases flammable gases.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P370+P378-In case of fire: Use to extinguish.
  • P422-Store contents under .
  • P501-Dispose of contents/container to .

SDS