missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Powder, -40+325 mesh, 99.8% (Metals basis), Alfa Aesar™

Brand:  Alfa Aesar 000010.A3

Code : NEW

Additional Details : CAS Number : 7429-90-5 Weight : 2.00000kg Transport : UN number : 1396 Chem class : 1 Pack group : II

Quantity 2kg

 View more versions of this product

Product Code. 11360639

  • £116.80 / 2kg


Estimated Shipment date
from Supplier 22-05-2018
Add to basket

Specifications

Chemical Identifiers

Aluminum
Powder
MFCD00134029
7429-90-5
99.8% (Metals basis)

Specifications

Aluminum
7429-90-5
400°C (752°F)
99.8% (Metals basis)
Odorless
Silver-gray
2kg
Silver Gray powder
Powder
MFCD00134029
Moisture sensitive
Videos
SDS