missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Silicon powder, -325 mesh, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4824592
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
11145-27-0
AlSi
55.067
MFCD00143595
CSDREXVUYHZDNP-UHFFFAOYSA-N
silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic
9793685
aluminum;silicon
[Al].[Si]
This item is not returnable. View return policy
11145-27-0
AlSi
55.067
MFCD00143595
CSDREXVUYHZDNP-UHFFFAOYSA-N
silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic
9793685
aluminum;silicon
[Al].[Si]

Aluminum Silicon powder is a reducing agent which is used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Appearance Silver Gray powder
Assay Percent Range 99% (Metals basis)
Quantity 100 g
Solubility Information Insoluble in water.
Mesh Size -325 Mesh
Odor Odorless
Form Powder
Chemical Name or Material Aluminum

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'