missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Powder, -100+325 mesh, 99.97% (Metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Silver to Gray |
Appearance | Silver Gray powder |
Assay Percent Range | 99.97% (Metals basis) |
Quantity | 5 g |
Ignition Point | 400°C (752°F) |
Mesh Size | -100+325 Mesh |
Odor | Odorless |
Sensitivity | Water reactive |
Form | Powder |
Chemical Name or Material | Aluminum |
Safety and Handling
Product Identifier
- Aluminum powder
- Danger
- Pyrophoric solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 2
- H250-Catches fire spontaneously if exposed to air.
- H261-In contact with water releases flammable gases.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P222-Do not allow contact with air.
- P231+P232-Handle under inert gas. Protect from moisture.
- P370+P378-In case of fire: Use to extinguish.
- P422-Store contents under .
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'