missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Potassium Sulfate, 12-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5390674
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
7784-24-9
AlH24KO20S2
474.372
GNHOJBNSNUXZQA-UHFFFAOYSA-J
kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate
62667
CHEBI:86465
aluminum;potassium;disulfate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]
This item is not returnable. View return policy
7784-24-9
AlH24KO20S2
474.372
GNHOJBNSNUXZQA-UHFFFAOYSA-J
kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate
62667
CHEBI:86465
aluminum;potassium;disulfate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]

High quality chemicals for laboratory and specialized industrial use

Physical Form Crystals
Quantity 500 g
Formula Weight 474.39
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.73kg
Chemical Name or Material Aluminum Potassium Sulfate, 12-Hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'