Learn More
Nickel Electro™ Aluminum Plate Sandbath Kit for use with HP1-1 Hotplates
Nickel Electro™ Aluminum Plate Sandbath Kit converts flat top HP1-1 hotplates into sandbath for immersion of small samples.
Brand: Nickel Electro™ SB1-1
Code : 03
Additional Details : Weight : 0.20000kg
Description
- Aluminum
- Used to covert hotplates into sand baths
Specifications
27 mm |