missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Aluminum plate, alloy 6061, 7.94mm (0.31 in.) thick

Brand:  Thermo Scientific Alfa Aesar 042127.RF

Aluminum plate, alloy 6061, 7.94mm (0.31 in.) thick

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15431787

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.