Learn More
Aluminum plate, alloy 6061, 3.18mm (0.125in) thick, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 045232.GT
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Aluminum plate, alloy 6061 is commonly used for structural components, frames, brackets, jigs, fixtures, base plates, machine parts, hydraulic valve bodies, valves parts, fuse parts, gears, worm gears, rectifier parts, fasteners, truck and marine components, marine fittings and hardware, electrical fittings and connectors, hinge pins, magneto parts, appliance fittings, camera lens mounts, etc. 6061 is used for heavy duty structures requiring good strength-to-weight ratio with good corrosion resistance. 6061 is easily cold worked and formed in the annealed condition. Cutting, stamping, bending, deep drawing, drilling, tapping, etc. are all readily accomplished using standard methods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Aluminum plate, Alloy 6061 | |
MFCD00134029 | |
Insoluble in water. | |
Plate |
3.18mm (0.125 in.) thick | |
150 x 150 mm | |
Odorless |
RUO – Research Use Only