missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum plate, alloy 6061, 2.3mm (0.09 in.) thick

Brand:  Alfa Aesar™ 045193.HB

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 300x300mm

 View more versions of this product

Product Code. 15432197

  • £48.50 / Each


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

Aluminum plate, Alloy 6061
2.3mm (0.09 in.) thick
MFCD00134029

Specifications

Aluminum plate, Alloy 6061
2.3mm (0.09 in.) thick
MFCD00134029
300x300mm
Plate
Odorless
Videos
SDS