missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum oxide, for TLC, with gypsum

For TLC

Brand:  Alfa Aesar™ 046113.36

Code : NEW

Additional Details : CAS Number : 1344-28-1 Weight : 0.50000kg

Quantity 500g

 View more versions of this product

Product Code. 15452617

  • £56.80 / 500g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

Aluminum oxide
With gypsum
101.96
14,356
1344-28-1
Al2O3
MFCD00003424

Specifications

Aluminum oxide
1344-28-1
Al2O3
MFCD00003424
Odorless
1.765
White
500g
With gypsum
Powder
101.96
14,356
5 to 15 Micron APS
2980°C
2045°C
Videos
SDS