missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum oxide, super activated, basic, Grade I

Brand:  Alfa Aesar™ 045886.A1

Code : NEW

Additional Details : CAS Number : 1344-28-1 Weight : 1.00000kg

Quantity 1kg

 View more versions of this product

Product Code. 15402537

  • £86.09 / 1kg

Estimated Shipment: 01-09-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
[O--].[O--].[O--].[Al+3].[Al+3]
Al2O3
MFCD00003424
aluminium oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
dialuminum;oxygen(2-)

Specifications

Aluminum oxide
1344-28-1
Al2O3
14,356
Insoluble in water,ethanol and diethyl ether.
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
101.96
Grade I
50 to 200 Micron APS
2980°C
2045°C
Super activated, basic
Al2O3
MFCD00003424
aluminium oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
50-200 Micron APS Powder
Odorless
1.765
White
1kg
Videos
SDS Documents
Certificates