Learn More
Aluminum oxide, super activated, acidic, Grade I, Thermo Scientific Chemicals
Description
Aluminum oxide is an inorganic compound used as a grinding or blending agent. In addition, Aluminum oxide can be essential in protein extraction as a component in the preparation of tissues. Aluminum oxide is a source of aluminum in reactions, an abrasive agent, and as a refractory material. Also Aluminum oxide can act as a catalyst in a variety of reactions such as the Claus process and in the dehydration of alcohols to alkenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 2980°C |
Melting Point | 2045°C |
Color | White |
Physical Form | Powder |
Quantity | 1 kg |
Linear Formula | Al2O3 |
Merck Index | 14,356 |
Solubility Information | Insoluble in water. |
IUPAC Name | dialuminum;oxygen(2-) |
Formula Weight | 101.96 |
Show More |
RUO – Research Use Only