missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Aluminum Oxide Substrate, 12.7 x 12.7 x 0.5mm, Polished One Side, R Plane, Alfa Aesar™
Brand: Alfa Aesar™ 043888.KT
Code : NEW
Additional Details : Weight : 0.01000kg
Quantity | 1 ea. |
---|
Chemical Identifiers
Aluminum Oxide Substrate, R Plane | |
Polished One Side | |
Al2O3 | |
MFCD00003424 |
Specifications
Aluminum Oxide Substrate, R Plane | |
Plate | |
MFCD00003424 | |
Off-white |
Polished One Side | |
Al2O3 | |
Odorless | |
1 ea. |