missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Aluminum oxide sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.99% (metals basis)
Brand: Alfa Aesar™ 041044.KS
Code : NEW
Additional Details : Weight : 0.01000kg
Specifications
Aluminum oxide sputtering target | |
99.99% | |
101.96 | |
Odorless |
76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick | |
(metals basis) | |
MFCD00003424 | |
1 each |