missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum oxide, Puriss., ≥98% Al2O3 basis, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
25 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Boiling Point | 2,980°C |
Melting Point | 2040°C (literature) |
CAS Max % | 100.0000% |
Quantity | 25 kg |
Linear Formula | Al2O3 |
UN Number | NONH for all modes of transport |
Formula Weight | 101.96 |
Percent Purity | >98% |
Grade | puriss. |
Packaging | Paper / Poly Bag |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'