missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, neutral, HPLC Flash Grade, Thermo Scientific Chemicals

Catalog Number p-7080026
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
9989226
[O--].[O--].[O--].[Al+3].[Al+3]
This item is not returnable. View return policy
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
9989226
[O--].[O--].[O--].[Al+3].[Al+3]

Aluminum oxide is an inorganic compound used as a grinding or blending agent. In addition, Aluminum oxide can be essential in protein extraction as a component in the preparation of tissues. Aluminum oxide is a source of aluminum in reactions, an abrasive agent, and as a refractory material. Also Aluminum oxide can act as a catalyst in a variety of reactions such as the Claus process and in the dehydration of alcohols to alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 2980°C
Melting Point 2045°C
Color White
Physical Form Powder
Quantity 250 g
Linear Formula Al2O3
Merck Index 14,356
Solubility Information Insoluble in water.
IUPAC Name dialuminium(3+) trioxidandiide
Formula Weight 101.96
Grade HPLC Flash
Odor Odorless
Refractive Index 1.765
Chemical Name or Material Aluminum oxide, Neutral
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'