missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, gamma-phase, 99.997% (metals basis), Alfa Aesar™

Brand:  Alfa Aesar 042576.22

Code : NEW

Additional Details : CAS Number : 1344-28-1 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11339188

  • £62.65 / 100g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

Aluminum oxide
gamma-phase
101.96
14,356
1344-28-1
Al2O3
MFCD00003424

Specifications

Aluminum oxide
1344-28-1
99.997%
Al2O3
MFCD00003424
Odorless
2980°C
100g
gamma-phase
110μm Powder
(metals basis)
101.96
14,356
60m2/g
2045°C
Videos
SDS