Learn More
Alfa Aesar™ Aluminum oxide, gamma-phase, 96+% incl. 3% C
Brand: Alfa Aesar™ 012867.14
Code : NEW
Additional Details : CAS Number : 1344-28-1 Weight : 0.02500kg
Quantity | 25g |
---|
Chemical Identifiers
Aluminum oxide | |
gamma-phase | |
MFCD00003424 |
1344-28-1 | |
101.96 | |
14,356 |
Specifications
Aluminum oxide | |
1344-28-1 | |
≥96% | |
MFCD00003424 | |
incl. 3% C | |
Odorless | |
175m2/g |
gamma-phase | |
Tablets | |
101.96 | |
14,356 | |
3.5g/mL to 3.9g/mL | |
3.2mm | |
25g |