missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum oxide, gamma-phase, 96+% incl. 3% C

Brand:  Alfa Aesar™ 012867.22

Code : NEW

Additional Details : CAS Number : 1344-28-1 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11357597

  • £65.40 / 100g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

Aluminum oxide
gamma-phase
MFCD00003424
1344-28-1
101.96
14,356

Specifications

Aluminum oxide
1344-28-1
≥96%
MFCD00003424
incl. 3% C
Odorless
175m2/g
gamma-phase
Tablets
101.96
14,356
3.5g/mL to 3.9g/mL
3.2mm
100g
Videos
SDS