missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Oxide, Catalyst Support, Intermediate Surface Area, Med pore diam 1.2 micron, Alfa Aesar

Brand:  Alfa Aesar 044693.A1

Code : NEW

Additional Details : Weight : 1.00000kg

Quantity 1kg

 View more versions of this product

Product Code. 11449910

  • £107.70 / 1kg


Estimated Shipment date
from Supplier 24-05-2018
Add to basket

Specifications

Chemical Identifiers

Aluminum Oxide
Intermediate Surface Area
MFCD00003424

Specifications

Aluminum Oxide
1344-28-1
MFCD00003424
3.2mm (0.125 in.)
Intermediate Surface Area
White pellets
Odorless
1kg
Videos
SDS