Learn More
Aluminum oxide, catalyst support, high surface area, 1/4in. Rings, Alfa Aesar™
S.A. 150 m2/g, medium pore diameter 130Å, total pore volume 0.77cc/g
Brand:  Alfa Aesar 043858.A7
Code : NEW
Additional Details : Weight : 5.00000kg
Quantity 5kg
 View more versions of this product
Product Code. 11340538
  • £519.60 / 5kg
Estimated Shipment date
from Supplier 12-06-2018
Add to basket
Specifications
Chemical Identifiers
Aluminum oxide
catalyst support, high surface area, 1/4in. Rings
MFCD00003424
Specifications
Aluminum oxide
1344-28-1
5kg
catalyst support, high surface area, 1/4in. Rings
MFCD00003424