missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum oxide, acidic, for TLC

For TLC

Brand:  Alfa Aesar™ 046099.22

Code : NEW

Additional Details : CAS Number : 1344-28-1 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15402617

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Aluminum oxide
Acidic
Al2O3
MFCD00003424
1344-28-1
Al2O3
101.96
14,356

Specifications

Aluminum oxide
1344-28-1
Al2O3
101.96
14,356
5 to 15 Micron APS
2980°C
2045°C
Acidic
Powder
Al2O3
MFCD00003424
Odorless
1.765
White
100g
Videos
SDS