missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum oxide, 99%, Thermo Scientific™
Click to view available options
Quantity:
500g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quantity | 500g |
Merck Index | 14,356 |
Solubility Information | Insoluble in water. |
Formula Weight | 101.96 |
Percent Purity | 99% |
Odor | Odorless |
Refractive Index | 1.768 |
Density | 3.965 g/mL |
Chemical Name or Material | Aluminum oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'