missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Nitride Paste, Alfa Aesar™

Brand:  Alfa Aesar 043959.14

Code : NEW

Additional Details : Weight : 0.02500kg


Quantity 25g

 View more versions of this product

Product Code. 11325008

  • £36.10 / 25g


Estimated Shipment date
from Supplier 26-04-2018
Add to basket


Specifications

Chemical Identifiers

Aluminium Nitride
Air set in 1-4 hours, heat cure in 2 hours at 200°C
AlN
MFCD00003429

Specifications

Aluminium Nitride
AlN
MFCD00003429
Gray
Air set in 1-4 hours, heat cure in 2 hours at 200°C
Gray paste
Odorless
25g
Videos
SDS