missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Nitride Paste, Alfa Aesar™

Brand:  Alfa Aesar 043959.22

Code : NEW

Additional Details : Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11335008

  • £113.30 / 100g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Aluminium Nitride
Air set in 1-4 hours, heat cure in 2 hours at 200°C
AlN
MFCD00003429

Specifications

Aluminium Nitride
AlN
MFCD00003429
Gray
Air set in 1-4 hours, heat cure in 2 hours at 200°C
Gray paste
Odorless
100g
Videos
SDS