missing translation for 'onlineSavingsMsg'
Learn More

Aluminum-nickel, Raney-type Alloy, Powder Al-Ni 50/50, ACROS Organics™

Brand:  Acros Organics 206235000

Code : 29

Additional Details : CAS Number : 12635-29-9 Weight : 0.55000kg Transport : UN number : 3208 Chem class : 1 Pack group : II


Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 10501081

  • £64.20 / 500g


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

Aluminium-nickel
12635-29-9
Al Ni
85.68

Specifications

47.0 to 52.0%
3.4600g/mL
1350.0°C
12635-29-9
Glass bottle
Al Ni
01,723; 02,293; 05,570; 06,502; 07,312; 01,723; 02,293; 05,570; 06,502; 07,312; 08,433; 09,405; 10,339; 11,457; 12,422; 13,265; 14,270; 15,278; 17,296
Aluminium-nickel
Gray to gray-black
500g
48.0 to 53.0%
Powder
85.68
Videos
SDS