missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Aluminum Magnesium plate, alloy 5052, 3.18mm (0.125 in.) thick

Brand:  Thermo Scientific Alfa Aesar 042124.RF

Aluminum Magnesium plate, alloy 5052, 3.18mm (0.125 in.) thick

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15446435

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.