missing translation for 'onlineSavingsMsg'
Learn More

Aluminum lanthanum isopropoxide, 99% (metals basis), 7% w/v in isopropanol, Thermo Scientific™

Catalog Number p-7051462
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
33939-97-8
MFCD00143591
Lanthanum aluminum isopropoxide
This item is not returnable. View return policy
33939-97-8
MFCD00143591
Lanthanum aluminum isopropoxide

Thin films of metal oxides

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 208°C (0.5 mmHg)
Quantity 25 mL
UN Number UN1219
Sensitivity Moisture sensitive
Formula Weight 928.24
Percent Purity 99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Aluminum lanthanum isopropoxide
compliance-icons compliance-icons
Product Identifier
  • Aluminum lanthanum isopropoxide 7% w/v in isopropanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'