missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum lactate, 90-95%, contains ^=5% water, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
It is used as a skin moisturizer lotion to treat dry, scaly, itchy skin, reagent in biological activity studies of aluminum and in sol-gel processing. Also used in fire foam. Aluminum L-Lactate has been used to inject aluminum into rabbits. Aluminum L-Lactate has been shown to decrease human enamel dissolution in a pH 4 acetate buffer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 100 g |
Merck Index | 14,348 |
Solubility Information | Very soluble in cold water |
Formula Weight | 294.2 |
Percent Purity | 90 to 95% |
Concentration or Composition (by Analyte or Components) | Contains ≈5% water |
Odor | Odorless |
Chemical Name or Material | Aluminum lactate |
Safety and Handling
Product Identifier
- Aluminum lactate
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'