missing translation for 'onlineSavingsMsg'
Learn More

Aluminum lactate, 90-95%, contains ^=5% water, Thermo Scientific Chemicals

Catalog Number p-7050618
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
18917-91-4
C9H15AlO9
294.19
MFCD20529175
VXYADVIJALMOEQ-UHFFFAOYNA-K
bis[(2-hydroxypropanoyl)oxy]alumanyl 2-hydroxypropanoate
CC(O)C(=O)O[Al](OC(=O)C(C)O)OC(=O)C(C)O
This item is not returnable. View return policy
18917-91-4
C9H15AlO9
294.19
MFCD20529175
VXYADVIJALMOEQ-UHFFFAOYNA-K
bis[(2-hydroxypropanoyl)oxy]alumanyl 2-hydroxypropanoate
CC(O)C(=O)O[Al](OC(=O)C(C)O)OC(=O)C(C)O

It is used as a skin moisturizer lotion to treat dry, scaly, itchy skin, reagent in biological activity studies of aluminum and in sol-gel processing. Also used in fire foam. Aluminum L-Lactate has been used to inject aluminum into rabbits. Aluminum L-Lactate has been shown to decrease human enamel dissolution in a pH 4 acetate buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 100 g
Merck Index 14,348
Solubility Information Very soluble in cold water
Formula Weight 294.2
Percent Purity 90 to 95%
Concentration or Composition (by Analyte or Components) Contains ≈5% water
Odor Odorless
Chemical Name or Material Aluminum lactate
compliance-icons
Product Identifier
  • Aluminum lactate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'