missing translation for 'onlineSavingsMsg'
Learn More

Aluminum isopropoxide, 98+%, Thermo Scientific Chemicals

Catalog Number p-7049325
29.66 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
555-31-7
C9H21AlO3
204.246
MFCD00008870
SMZOGRDCAXLAAR-UHFFFAOYSA-N
aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate
11143
aluminum;propan-2-olate
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
555-31-7
C9H21AlO3
204.246
MFCD00008870
SMZOGRDCAXLAAR-UHFFFAOYSA-N
aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate
11143
aluminum;propan-2-olate
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3]

Reagent for the reduction of carbonyl compounds

Reagent for the reduction of carbonyl compounds. Used in alcoholysis and ester exchange reactions. Also used in synthesis of higher alkoxides, chelates, and acrylates. Aluminum isopropoxide is a useful reagent in the synthesis of alkoxides, chelates and acrylates. It acts as an intermediate of pharmaceutical products like isophytol, testosterone, progesterone and ethisterone. Also used as the material of aluminate coupling agent. It is also used in alcoholysis and ester exchange reactions. In Meerwein-Ponndorf-Verley (MPV) reduction, it is used as a catalyst for the reduction of ketones and aldehydes to their corresponding alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Reagent for the reduction of carbonyl compounds
  • Used in alcoholysis and ester exchange reactions
  • Also used in synthesis of higher alkoxides, chelates, and acrylates
TRUSTED_SUSTAINABILITY
Melting Point 131°C to 135°C
Density 1.035
Boiling Point 135°C (10 mm)
Assay Percent Range 98+%
Refractive Index 1.0346
Quantity 1 kg
UN Number UN3181
Sensitivity Moisture sensitive
Merck Index 14,347
Solubility Information Soluble in alcohol,benzene,and toluene; Decomposes in water;Soluble in ethanol,isopropanol,benzene,toluene,chloroform,carbon tetrachloride and petroleum hydrocarbons.
Formula Weight 204.25
Percent Purity ≥98%
Physical Form Granules
Chemical Name or Material Aluminum isopropoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'