missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum foil, alloy 6061, 1.6mm (0.063 in.) thick

Brand:  Alfa Aesar™ 045212.HB

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 300x300mm

Product Code. 15452207

  • £33.30 / Each


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Specifications

Aluminum foil, Alloy 6061
1.6mm (0.063 in.) thick
MFCD00134029
300x300mm
Foil
Odorless
Videos
SDS