missing translation for 'onlineSavingsMsg'
Learn More

Aluminum foil, 1.0mm (0.04 in.) thick, annealed, 99% (metals basis), Si and Fe (combined) typically 1%, Thermo Scientific Chemicals

Catalog Number p-7049371
Click to view available options
Quantity:
45 x 60 cm
45 x 240 cm
5 x 45 x 240 cm
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
aluminum
[Al]
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
aluminum
[Al]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 x 45 x 240 cm
Percent Purity 99%
Concentration or Composition (by Analyte or Components) Si and Fe (combined) typically 1%
Odor Odorless
Weight ≈729g/45x60cm
Form Foil
Assay (metals basis)
Chemical Name or Material Aluminum foil, 1.0mm (0.04 in.) thick, Annealed, Temper: soft

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'