missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum fluoride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Aluminum fluoride is used in the smelting process to lower the melting point of electrolytes, production of aluminum silicates, refining of aluminum scarp, production of specialty refractory products, and in glass industry as filler. It is used as flux ingredient to remove magnesium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1291°C (sublimation) |
Physical Form | Powder |
Quantity | 10 g |
Merck Index | 14,339 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 83.98 |
Percent Purity | 99.99% |
Sensitivity | Hygroscopic |
Density | 2.882 g/mL |
Assay | (metals basis) |
Show More |
Safety and Handling
Product Identifier
- Aluminum fluoride, anhydrous
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'