missing translation for 'onlineSavingsMsg'
Learn More

Aluminum fluoride, anhydrous, 99+%, Thermo Scientific Chemicals

Catalog Number p-7048796
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy
7784-18-1
MFCD00003426
This item is not returnable. View return policy
7784-18-1
MFCD00003426

In ceramics, as flux in metallurgy, in aluminum manufacture, as inhibitor of fermentation, as catalyst in organic reactionsAluminum fluoride is used in the smelting process to lower the melting point of electrolytes, production of aluminum silicates, refining of aluminum scarp, production of specialty refractory products, and in glass industry as filler. It is used as flux ingredient to remove magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In ceramics, as flux in metallurgy, in aluminum manufacture, as inhibitor of fermentation, as catalyst in organic reactions
TRUSTED_SUSTAINABILITY
Melting Point 1291°C (sublimation)
Physical Form Powder
Quantity 1 g
Merck Index 14,339
Solubility Information Practically insoluble in water; Sparingly soluble in acids and alkalies.
Formula Weight 83.98
Percent Purity ≥99%
Sensitivity Hygroscopic
Density 2.882 g/mL
Chemical Name or Material Aluminum fluoride, Anhydrous
compliance-icons compliance-icons
Product Identifier
  • Aluminum fluoride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'