missing translation for 'onlineSavingsMsg'
Learn More

Aluminum flake, APS 11 micron, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog Number 15446685
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
[Al]
This item is not returnable. View return policy
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
[Al]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Appearance Silver Gray flakes
Density 2.7g/cm3
Assay Percent Range 99.7% (Metals basis)
Quantity 250 g
Ignition Point 400°C (752°F)
IUPAC Name aluminum
Odor Odorless
Sensitivity Water reactive
Form Flake
Chemical Name or Material Aluminum
Show More Show Less
compliance-icons
Product Identifier
  • Aluminum flake
Signal Word
  • Danger
Hazard Category
  • Pyrophoric solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P370+P378-In case of fire: Use to extinguish.
  • P422-Store contents under .
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'