missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Aluminum Fixed Angle Rotors

Use these rotors for a variety of applications. Eppendorf™ Aluminum Fixed Angle Rotors are ideal for molecular and biological laboratories.

Brand:  Eppendorf™ 5820715006

Code : 99

Additional Details : Weight : 3.50000kg

 View more versions of this product

Product Code. 10733912

  • £1655.00 / Each


Estimated Shipment
17-05-2018
Add to basket

Description

Description

  • Choice of four different rotors
  • Standard rotor can accommodate 1.5mL/2.0mL tubes
  • PCR* strips and kits/spin columns can be centrifuged in the corresponding rotor
Specifications

Specifications

Aluminum
6 x 15/50mL
12,100rpm
Fixed Angle Rotor
20,130 x g
Videos