Learn More
Eppendorf™ Aluminum Fixed Angle Rotors
Use these rotors for a variety of applications. Eppendorf™ Aluminum Fixed Angle Rotors are ideal for molecular and biological laboratories.
Brand:  Eppendorf™ 5490025008
Code : 99
Additional Details : Weight : 1.26000kg
 View more versions of this product
Product Code. 10338321
  • £468.35 / Each
Estimated Shipment
22-02-2018
Add to basket
Description
Description
  • Choice of four different rotors
  • Standard rotor can accommodate 1.5mL/2.0mL tubes
  • PCR* strips and kits/spin columns can be centrifuged in the corresponding rotor
Specifications
Specifications
Aluminum/Polycarbonate
Fixed Angle Rotor
30 x 1.5/2mL
Transparent Polycarbonate Lid
Videos
Documentation View accessory products