Learn More
Eppendorf™ Aluminum Fixed Angle Rotors
Use these rotors for a variety of applications. Eppendorf™ Aluminum Fixed Angle Rotors are ideal for molecular and biological laboratories.
Brand: Eppendorf™ 5490015002
Code : 99
Additional Details : Weight : 1.25000kg
Description
- Choice of four different rotors
- Standard rotor can accommodate 1.5mL/2.0mL tubes
- PCR* strips and kits/spin columns can be centrifuged in the corresponding rotor
Specifications
Aluminum | |
30 x 1.5/2mL | |
— |
Fixed Angle Rotor | |
20,800 x g | |
Aluminum Lid |