missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Aluminum Fixed Angle Rotors

Use these rotors for a variety of applications. Eppendorf™ Aluminum Fixed Angle Rotors are ideal for molecular and biological laboratories.

Brand:  Eppendorf™ 5425737009

Code : 99

Additional Details : Weight : 1.01700kg

 View more versions of this product

Product Code. 10273932

  • £418.20 / Each


Estimated Shipment
14-02-2018
Add to basket

Description

Description

  • Choice of four different rotors
  • Standard rotor can accommodate 1.5mL/2.0mL tubes
  • PCR* strips and kits/spin columns can be centrifuged in the corresponding rotor
Specifications

Specifications

Aluminum
24 x 1.5/2mL
13,200rpm
Fixed Angle Rotor
16,100 x g
Aerosol rotor lid
Videos
Documentation View accessory products