Learn More
Eppendorf™ Aluminum Fixed Angle Rotors
Use these rotors for a variety of applications. Eppendorf™ Aluminum Fixed Angle Rotors are ideal for molecular and biological laboratories.
Brand: Eppendorf™ 5427714006
Code : 99
Additional Details : Weight : 1.00000kg
Includes: Lid (see details for type)
Description
Aerosol-tight rotors (05-401-503, 05-400-023 and 05-401-505) feature a distinct red rotor lid screw and red ring inside the rotor for easy identification. Tube adapters are available separately.
Standard 30-place Rotors
- Capacity: 30 × 1.5/2.0mL Molecular-grade Max. Speed/RCF: 14,000rpm/20,8170xG
- Available with aerosol-tight lid for use with hazardous samples (05-401-503), or with plastic lid for basic applications (05-400-019)
- PTFE-coated for high chemical resistance and easy cleaning
- PTFE-coated for high chemical resistance and easy cleaning
- Capacity: 24 × 1.5/2.0mL tubes
- Max. Speed/RCF: 17,500rpm/30,1300xG
- Includes aerosol-tight lid for hazardous sample work
- Check tube stability before use; Eppendorf 1.5mL Flex-Tube™ or Safe-Lock™ Tubes recommended
- Safely spins two standard microplates
- Max. Speed/RCF: 4680rpm/22,040xG
- Loading height of 29mm is suitable for all types of MTP or PCR plates
- Windshielded design and drop-on lid ensure virtually silent centrifugation
15mL/50mL Conical Tube Rotor
- Capacity: 6 × 15mL or 6 × 50mL conical tubes
- Max. Speed/RCF: 7830rpm/7,197xG
- Adapters for 15 and 50mL conical tubes included
- 50mL adapter also accommodates 50mL skirted conical tubes
24-place Spin Column Kit Rotor
- Capacity: 24 × spin columns or 24 × 1.5/2.0mL tubes
- Max. Speed/RCF: 13,200rpm/19,090xG
- Unique high rim design supports open tube lids during centrifugation; lids will not tear off
- Aerosol-tight lid for hazardous samples
PCR Strip Rotor
- Capacity: 8 × 8 PCR tube strips or 64 × 0.2mL PCR tubes
- Max. Speed/RCF: 11,800rpm/13,5480xG
- Includes removable adapters that can be used as a tube rack for pipetting
Cryotube Rotor
- Ideal for tissue culture labs that need to spin down samples before cryo conservation
- Accepts up to 18 cryo vials
- Max. Speed/RCF: 8900rpm/83,200xG
- Includes adapters to accommodate tubes with 16.9 or 13.4mm diameter
Certifications
Aerosol-tight rotors are tested and certified by the Health Protection Agency (Porton Down, UK)
Specifications
PTFE Coated | |
64 x 0.2 or 8 x 8 mL | |
11,800 rpm | |
5430 Microcentrifuge | |
Screw-on lid |
Rotor | |
13,543 x g | |
8.7 cm | |
Fixed |