missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum ethoxide, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Aluminum ethoxide is mainly used in industrial settings. It is used as a reducing agent for aldehyde, ketones, polymerization catalyst. Used in the sol-gel process preparation of high purity aluminum sesquioxide. It also used as a reducing reagent, for carbonyl compounds that restore to alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 154°C to 159°C |
Color | White |
Density | 1.142 g/mL |
Boiling Point | 320°C |
Linear Formula | Al(OC2H5)3 |
Quantity | 10 g |
UN Number | UN2925 |
Sensitivity | Moisture sensitive |
Merck Index | 14,338 |
Solubility Information | Soluble in hot xylene,cholorobenzene. Reacts with water. |
Show More |
Safety and Handling
Product Identifier
- Aluminum ethoxide
- Danger
- Flammable solid Category 1
- Skin corrosion/irritation Category 1 B
- H228-Flammable solid.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'