missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum ethoxide, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Aluminum ethoxide is mainly used in industrial settings. It is used as a reducing agent for aldehyde, ketones, polymerization catalyst. Used in the sol-gel process preparation of high purity aluminum sesquioxide. It also used as a reducing reagent, for carbonyl compounds that restore to alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 154°C to 159°C |
Color | White |
Density | 1.142 g/mL |
Boiling Point | 320°C |
Linear Formula | Al(OC2H5)3 |
Quantity | 50 g |
UN Number | UN2925 |
Sensitivity | Moisture sensitive |
Merck Index | 14,338 |
Solubility Information | Soluble in hot xylene,cholorobenzene. Reacts with water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'