missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum dihydrogen phosphate, 50% w/w aqueous solution, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
5 kg
Description
Aluminum dihydrogen phosphate is used as intermediates and solid separation agents. It is also used in petroleum, chemical industry, shipbuilding, space technology, electric industry, high-temperature kilns, heat treatment resistance furnaces and electric insulation. Further, it is used as inorganic coating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 1 kg |
Assay Percent Range | 50% w/w aqueous solution |
Solubility Information | Soluble in water and ethanol. Insoluble in acetone. |
Formula Weight | 317.94 |
Appearance | Colorless liquid |
Density | 1.5 |
Chemical Name or Material | Aluminum dihydrogen phosphate |
Safety and Handling
Product Identifier
- Aluminum dihydrogen phosphate, 50% w/w aq. Soln
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Aluminum dihydrogenphosphate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'