missing translation for 'onlineSavingsMsg'
Learn More

Aluminum di(sec-butoxide)acetoacetic ester chelate, tech., Al 8.4% min, Thermo Scientific Chemicals

Catalog Number 15417515
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price

 

Aluminum di(sec-butoxide)acetoacetic ester chelate is used in addition with lithium acetate and titanium(IV) ethoxide to prepare precursor sol to prepare Li1.3Al0.3Ti1.7(PO4)3 thin films on glass substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 24772-51-8
Density 1.03 g/mL
Flash Point 76°C (169°F)
Odor Alcohol-like
Molecular Formula C14H27AlO5
MDL Number MFCD00084762
Quantity 500 g
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 302.3
Grade Technical
Physical Form Liquid
Chemical Name or Material Aluminum di (sec-butoxide)acetoacetic ester chelate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'