missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Aluminum Copper square bar, alloy 2024, 2.54cm (1.0in) x 2.54cm (1.0in)

Brand:  Thermo Scientific Alfa Aesar 042106.BT

Aluminum Copper square bar, alloy 2024, 2.54cm (1.0in) x 2.54cm (1.0in)

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11359757

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.