missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum Copper square bar, alloy 2024, 2.54cm (1.0in) x 2.54cm (1.0in)

Brand:  Alfa Aesar™ 042106.RS

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 60cm

 View more versions of this product

Product Code. 11369757

  • £82.10 / 60cm


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Specifications

Chemical Identifiers

Aluminum Copper square bar, Alloy 2024
2.54cm (1.0 in.) x 2.54cm (1.0 in.)
MFCD00213972

Specifications

Al:Cu:Mg:Mn; 93.5:4.4:1.5:0.6 wt%
≈17.92g/cm
2.54cm (1.0 in.) x 2.54cm (1.0 in.)
MFCD00213972
Aluminum Copper square bar, Alloy 2024
60cm
Bar
Odorless
Videos
SDS