missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Aluminum Copper square bar, alloy 2024, 1.91cm (0.75in) x 1.91cm (0.75in)

Brand:  Thermo Scientific Alfa Aesar 042105.BT

Aluminum Copper square bar, alloy 2024, 1.91cm (0.75in) x 1.91cm (0.75in)

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11349757

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.