missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Aluminum Copper spheres, alloy 2017, 9.5mm (0.37 in.) dia.

Brand:  Thermo Scientific Alfa Aesar 042034.KG

Aluminum Copper spheres, alloy 2017, 9.5mm (0.37 in.) dia.

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11395969

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.